Assay Details
| Gene Symbol: LOC100385845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004322.2 | XP_009002570.2 | 1 - 2 | 76 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100385845| Gene Symbol: LOC100385845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004322.2 | XP_009002570.2 | 1 - 2 | 76 | 95 | |
Set Membership: |
|