Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747169.2 | XP_002747215.1 | 7 - 8 | 1179 | 97 | |
| XM_002747170.3 | XP_002747216.1 | 6 - 7 | 860 | 97 | ||
More Information
Set Membership: |
|

TBXT
Genomic Map