Assay Details
| Gene Symbol: SGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759645.4 | XP_002759691.1 | 3 - 4 | 520 | 72 | |
| XM_008983898.2 | XP_008982146.1 | 3 - 4 | 516 | 72 | ||
| XM_008983899.2 | XP_008982147.1 | 3 - 4 | 518 | 72 | ||
| XM_008983900.2 | XP_008982148.1 | 3 - 4 | 518 | 72 | ||
| XM_017965893.1 | XP_017821382.1 | 3 - 4 | 520 | 72 | ||
More Information
Set Membership: |
|

SGO1
Genomic Map