Assay Details
Gene Symbol: LOC100389516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003732570.3 | XP_003732618.1 | 1 | 619 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
