Assay Details
| Gene Symbol: LOC100386949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980930.1 | XP_008979178.1 | 1 - 2 | 306 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100386949| Gene Symbol: LOC100386949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980930.1 | XP_008979178.1 | 1 - 2 | 306 | 114 | |
Set Membership: |
|