Assay Details
| Gene Symbol: HHLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982522.2 | XP_008980770.1 | 3 - 4 | 526 | 104 | |
| XM_008982524.2 | XP_008980772.1 | 3 - 4 | 2349 | 104 | ||
More Information
Set Membership: |
|

HHLA2
Genomic Map