Assay Details
| Gene Symbol: LOC100411271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988633.2 | XP_008986881.2 | 5 - 6 | 1121 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100411271| Gene Symbol: LOC100411271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988633.2 | XP_008986881.2 | 5 - 6 | 1121 | 68 | |
Set Membership: |
|