Assay Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998758.2 | XP_008997006.2 | 2 - 3 | 787 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

G6PC2| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998758.2 | XP_008997006.2 | 2 - 3 | 787 | 71 | |
Set Membership: |
|