Assay Details
| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009151.2 | XP_009007399.1 | 4 - 5 | 1187 | 93 | |
| XM_017978717.1 | XP_017834206.1 | 4 - 5 | 1107 | 93 | ||
| XM_017978718.1 | XP_017834207.1 | 4 - 5 | 1187 | 93 | ||
More Information
Set Membership: |
|

ATF7IP2
Genomic Map