Assay Details
| Gene Symbol: LOC100396683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751982.3 | XP_002752028.1 | 12 - 13 | 1631 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100396683| Gene Symbol: LOC100396683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751982.3 | XP_002752028.1 | 12 - 13 | 1631 | 66 | |
Set Membership: |
|