Assay Details
| Gene Symbol: LOC100396683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751982.3 | XP_002752028.1 | 9 - 10 | 1326 | 89 | |
| XM_017975567.1 | XP_017831056.1 | 9 - 10 | 1326 | 89 | ||
More Information
Set Membership: |
|

LOC100396683
Genomic Map