Assay Details
| Gene Symbol: CRELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734888.3 | XP_003734936.1 | 7 - 8 | 835 | 58 | |
| XM_008982133.2 | XP_008980381.1 | 7 - 8 | 802 | 58 | ||
More Information
Set Membership: |
|

CRELD1
Genomic Map