Assay Details
| Gene Symbol: CNGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007587.2 | XP_009005835.2 | 4 - 5 | 1068 | 84 | |
| XM_017977864.1 | XP_017833353.1 | 4 - 5 | 1068 | 84 | ||
More Information
Set Membership: |
|

CNGA4
Genomic Map