Assay Details
| Gene Symbol: BEAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966759.1 | XP_017822248.1 | 2 - 3 | 236 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BEAN1| Gene Symbol: BEAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966759.1 | XP_017822248.1 | 2 - 3 | 236 | 118 | |
Set Membership: |
|