Assay Details
| Gene Symbol: BHMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967667.1 | XP_017823156.1 | 6 - 7 | 845 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BHMG1| Gene Symbol: BHMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967667.1 | XP_017823156.1 | 6 - 7 | 845 | 108 | |
Set Membership: |
|