Assay Details
| Gene Symbol: LOC103795618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976863.1 | XP_017832352.1 | 2 - 3 | 953 | 89 | |
| XM_017976864.1 | XP_017832353.1 | 1 - 2 | 811 | 89 | ||
| XM_017976865.1 | XP_017832354.1 | 2 - 3 | 953 | 89 | ||
| XM_017976866.1 | XP_017832355.1 | 2 - 3 | 953 | 89 | ||
More Information
Set Membership: |
|

LOC103795618
Genomic Map