Assay Details
| Gene Symbol: LOC103795618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976863.1 | XP_017832352.1 | 7 - 8 | 1550 | 80 | |
| XM_017976864.1 | XP_017832353.1 | 6 - 7 | 1408 | 80 | ||
More Information
Set Membership: |
|

LOC103795618
Genomic Map