Assay Details
| Gene Symbol: LAIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762481.3 | XP_002762527.1 | 3 - 4 | 462 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAIR1| Gene Symbol: LAIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762481.3 | XP_002762527.1 | 3 - 4 | 462 | 87 | |
Set Membership: |
|