Assay Details
Gene Symbol: ACSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003734495.2 | XP_003734543.1 | 5 - 6 | 892 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
