Assay Details
| Gene Symbol: LOC100393774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972865.1 | XP_017828354.1 | 8 - 9 | 1203 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100393774| Gene Symbol: LOC100393774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972865.1 | XP_017828354.1 | 8 - 9 | 1203 | 90 | |