Assay Details
| Gene Symbol: LOC103793577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973139.1 | XP_017828628.1 | 9 - 10 | 1417 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103793577| Gene Symbol: LOC103793577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973139.1 | XP_017828628.1 | 9 - 10 | 1417 | 117 | |
Set Membership: |
|