Assay Details
Gene Symbol: CD300C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748704.3 | XP_002748750.1 | 2 - 3 | 783 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
