Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732741.3 | XP_003732789.1 | 7 - 8 | 1159 | 74 | |
| XM_008995123.1 | XP_008993371.1 | 7 - 8 | 1431 | 74 | ||
| XM_008995124.2 | XP_008993372.1 | 6 - 7 | 1247 | 74 | ||
| XM_017971416.1 | XP_017826905.1 | 6 - 7 | 1031 | 74 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map