Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984673.2 | XP_008982921.1 | 4 - 5 | 906 | 84 | |
| XM_008984675.2 | XP_008982923.1 | 5 - 6 | 1183 | 84 | ||
| XM_017966378.1 | XP_017821867.1 | 3 - 4 | 555 | 84 | ||
| XM_017966380.1 | XP_017821869.1 | 3 - 4 | 616 | 84 | ||
More Information
Set Membership: |
|

CD1E
Genomic Map