Assay Details
| Gene Symbol: KCNH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996876.2 | XP_008995124.1 | 2 - 3 | 354 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNH6| Gene Symbol: KCNH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996876.2 | XP_008995124.1 | 2 - 3 | 354 | 74 | |
Set Membership: |
|