Assay Details
| Gene Symbol: C19H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966694.1 | XP_017822183.1 | 3 - 4 | 600 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19H1orf100| Gene Symbol: C19H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966694.1 | XP_017822183.1 | 3 - 4 | 600 | 101 | |
Set Membership: |
|