Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747389.3 | XP_002747435.1 | 1 - 2 | 434 | 69 | |
| XM_008995617.1 | XP_008993865.1 | 1 - 2 | 434 | 69 | ||
More Information
Set Membership: |
|

ANGPT4
Genomic Map