Assay Details
| Gene Symbol: AKR1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750008.4 | XP_002750054.1 | 2 - 3 | 339 | 73 | |
| XM_008999867.2 | XP_008998115.1 | 2 - 3 | 343 | 73 | ||
More Information
Set Membership: |
|

AKR1C4
Genomic Map