Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750623.4 | XP_002750669.1 | 2 - 3 | 285 | 73 | |
| XM_009000918.2 | XP_008999166.1 | 1 - 2 | 376 | 73 | ||
More Information
Set Membership: |
|

CDCA8
Genomic Map