Assay Details
| Gene Symbol: NKX1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008978865.2 | XP_008977113.1 | 1 - 2 | 212 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX1-2| Gene Symbol: NKX1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008978865.2 | XP_008977113.1 | 1 - 2 | 212 | 115 | |
Set Membership: |
|