Assay Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978345.1 | XP_017833834.1 | 3 - 4 | 871 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TESMIN| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978345.1 | XP_017833834.1 | 3 - 4 | 871 | 93 | |