Assay Details
| Gene Symbol: CBLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989072.2 | XP_008987320.1 | 1 | 1123 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBLL2| Gene Symbol: CBLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989072.2 | XP_008987320.1 | 1 | 1123 | 100 | |
Set Membership: |
|