Assay Details
| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745787.2 | XP_002745833.1 | 9 - 10 | 1212 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11F| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745787.2 | XP_002745833.1 | 9 - 10 | 1212 | 120 | |
Set Membership: |
|