Assay Details
| Gene Symbol: ISX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732130.3 | XP_003732178.2 | 1 - 2 | 1296 | 108 | |
| XM_017963286.1 | XP_017818775.1 | 2 - 3 | 1019 | 108 | ||
More Information
Set Membership: |
|

ISX
Genomic Map