Assay Details
| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998002.2 | XP_008996250.1 | 3 - 4 | 328 | 107 | |
| XM_008998003.2 | XP_008996251.1 | 3 - 4 | 321 | 107 | ||
| XM_008998004.2 | XP_008996252.1 | 3 - 4 | 321 | 107 | ||
More Information
Set Membership: |
|

EEF1AKMT1
Genomic Map