Assay Details
| Gene Symbol: PRODH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987766.2 | XP_008986014.1 | 4 - 5 | 700 | 59 | |
| XM_008987767.2 | XP_008986015.1 | 4 - 5 | 701 | 59 | ||
More Information
Set Membership: |
|

PRODH2
Genomic Map