Assay Details
| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 12 - 13 | 3630 | 94 | |
| XM_017976342.1 | XP_017831831.1 | 12 - 13 | 3628 | 94 | ||
More Information
Set Membership: |
|

LOC100406494
Genomic Map