Assay Details
| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 2 - 3 | 2360 | 78 | |
| XM_017976342.1 | XP_017831831.1 | 2 - 3 | 2361 | 78 | ||
More Information
Set Membership: |
|

LOC100406494
Genomic Map