Assay Details
| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 6 - 7 | 2832 | 86 | |
| XM_017976342.1 | XP_017831831.1 | 6 - 7 | 2833 | 86 | ||
More Information
Set Membership: |
|

LOC100406494
Genomic Map