Assay Details
| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 9 - 10 | 3165 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100406494| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 9 - 10 | 3165 | 66 | |
Set Membership: |
|