Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752097.4 | XP_002752143.1 | 9 - 10 | 1280 | 65 | |
| XM_009003229.2 | XP_009001477.1 | 8 - 9 | 1157 | 65 | ||
More Information
Set Membership: |
|

SLCO1B3
Genomic Map