Assay Details
| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987292.2 | XP_008985540.1 | 2 - 3 | 424 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPAN16| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987292.2 | XP_008985540.1 | 2 - 3 | 424 | 75 | |