Assay Details
| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987292.2 | XP_008985540.1 | 4 - 5 | 613 | 92 | |
| XM_017967799.1 | XP_017823288.1 | 3 - 4 | 538 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPAN16| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987292.2 | XP_008985540.1 | 4 - 5 | 613 | 92 | |
| XM_017967799.1 | XP_017823288.1 | 3 - 4 | 538 | 92 | ||