Assay Details
| Gene Symbol: PRKACG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995942.2 | XP_008994190.1 | 1 | 1923 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKACG| Gene Symbol: PRKACG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995942.2 | XP_008994190.1 | 1 | 1923 | 103 | |
Set Membership: |
|