Assay Details
| Gene Symbol: SSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746247.4 | XP_002746293.1 | 2 - 3 | 392 | 82 | |
| XM_003732595.2 | XP_003732643.1 | 2 - 3 | 392 | 82 | ||
| XM_008994005.2 | XP_008992253.1 | 2 - 3 | 392 | 82 | ||
| XM_008994006.2 | XP_008992254.1 | 2 - 3 | 392 | 82 | ||
| XM_008994007.2 | XP_008992255.1 | 2 - 3 | 374 | 82 | ||
| XM_008994008.2 | XP_008992256.1 | 2 - 3 | 374 | 82 | ||
More Information
Set Membership: |
|

SSR1
Genomic Map