Assay Details
| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756926.3 | XP_002756972.1 | 2 - 3 | 753 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2CB| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756926.3 | XP_002756972.1 | 2 - 3 | 753 | 86 | |
Set Membership: |
|