Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996162.2 | XP_008994410.1 | 7 - 8 | 1436 | 65 | |
| XM_017972128.1 | XP_017827617.1 | 7 - 8 | 1435 | 65 | ||
More Information
Set Membership: |
|

CTCFL
Genomic Map