Assay Details
| Gene Symbol: SPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004459.2 | XP_009002707.1 | 5 - 6 | 614 | 70 | |
| XM_009004460.2 | XP_009002708.1 | 5 - 6 | 565 | 70 | ||
| XM_017976315.1 | XP_017831804.1 | 6 - 7 | 574 | 70 | ||
| XM_017976316.1 | XP_017831805.1 | 5 - 6 | 611 | 70 | ||
More Information
Set Membership: |
|

SPIC
Genomic Map