Assay Details
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017967295.1 | XP_017822784.1 | 11 - 12 | 1375 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
