Assay Details
| Gene Symbol: ERICH6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971773.1 | XP_017827262.1 | 9 - 10 | 1931 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH6B| Gene Symbol: ERICH6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971773.1 | XP_017827262.1 | 9 - 10 | 1931 | 92 | |